Learn More
Tocris Bioscience™ ω-Agatoxin IVA
CaV2.1 blocker
Brand:  Tocris Bioscience™ 2799/10U
188.58 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18034192
  • 238.00€ / 10µg
Estimated Shipment: 03-01-2025
Description
ω-Agatoxin IVA is a selective blocker of P-type calcium channels (IC50 < 1 - 3 nM). Also inhibits N-type channels at micromolar concentrations.
Specifications
Specifications
95%
Store at -20°C