Learn More
Tocris Bioscience™ [D-p-Cl-Phe6,Leu17]-VIP
Selective VIP receptor antagonist
Brand:  Tocris Bioscience™ 3054/1
313.41 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18095082
  • 387.00€ / 1mg
Estimated Shipment: 03-01-2025
Description
[D-p-Cl-Phe6,Leu17]-VIP is a selective vasoactive intestinal peptide (VIP) receptor antagonist (IC50 = 125.8 nM). Displays no activity on glucagon, secretin or GRF receptors.