Learn More
Tocris Bioscience™ Metastin (human)
Potent, endogenous ligand for kisspeptin receptor
Brand:  Tocris Bioscience™ 1443/100U
Product Code. 18015362
  • 345.00€ / 100µg
Estimated Shipment: 14-03-2025
Description
Metastin (human) is a potent endogenous ligand of the kisspeptin receptor (KISS1, GPR54). Binds with high affinity to rat and human KISS1 receptors with Ki values of 1.80 and 1.45 nM respectively. Inhibits chemotaxis, invasion and metastasis of human melanomas and breast carcinomas. Stimulates gonadotropin secretion following i.c.v. administration.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
95%
Store at -20°C
100 μg
Product Title
Tocris Bioscience™ Metastin (human) > 100ug