missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TOE1 Mouse, anti-Human, Unconjugated, Clone: 2C10H7, Proteintech
Mouse Monoclonal Antibody
145.00€ - 410.00€
Specifications
Antigen | TOE1 |
---|---|
Clone | 2C10H7 |
Concentration | 1000 μg/mL |
Applications | Western Blot, ELISA |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17211486
|
Proteintech
67829-1-IG-20UL |
20 μL |
145.00€
20µL |
Estimated Shipment: 30-01-2025
Log in to see stock. |
|||||
17201486
|
Proteintech
67829-1-IG-150UL |
150 μL |
410.00€
150µL |
In Stock
Log in to see stock. |
|||||
Description
The TOE1 antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human TOE1. This antibody recognizes human antigen. The TOE1 antibody has been validated for the following applications: WB,ELISA analysis.
Specifications
TOE1 | |
1000 μg/mL | |
Monoclonal | |
Liquid | |
Human | |
Primary |
2C10H7 | |
Western Blot, ELISA | |
Unconjugated | |
Mouse | |
114034 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title