Learn More
Thermo Scientific™ TopVision Agarose PROMO
TopVision Agarose is highly purified agarose suitable for analytical and preparative electrophoresis of nucleic acids.
Brand:  Thermo Scientific™ R0492-RMD123
Product Code. 15659484
Description
Description
Thermo Scientific TopVision Agarose provides optimal concentration between 0.7 to 2% in all typical buffer systems. It is a highly purified agarose with very low EEO values certified by strict quality control test procedures.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Agarose Gel
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ TopVision Agarose PROMO >