missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Total and Free Cholesterol Assay Kit (Fluorometric)

Assay Kit (Fluorometric)

Brand:  Novus Biologicals™ NBP3-25935

Product Code. 18677920

  • 461.00€ / 1 set
Estimated Shipment: 23-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

Storage of components varies. See protocol for specific instructions.
96 Tests
Non-species specific
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers