Learn More
STEMCELL Technologies Total RNA Purification Kit
For purification of RNA from cells
Brand:  STEMCELL Technologies 79040
Encompass_Preferred
Product Code. 17129911
  • 534.60€ / 1 set
Estimated Shipment: 27-11-2024
Description
  • The Total RNA Purification Kit uses a membrane-based system to quickly purify intact total RNA from 1 x 10^2 to 5 x 10^6 cells without an organic extraction step, eliminating the need for additional purification steps
  • The Total RNA Purification Kit includes an on-column DNase I treatment to significantly reduce genomic DNA contamination and ensure the purified RNA is suitable for sensitive downstream applications
  • Intact total RNA can be isolated in as little as 30 minutes
  • The Total RNA Purification Kit is not recommended for the purification of small RNAs (< 200 nt), such as miRNA
Specifications
Specifications
50 Preps