Learn More
Gilac™ Tote Caddy
Tote Caddy is highly durable and suitable for universal use.
Brand:  Gilac™ G161438
 View more versions of this product
Product Code. 17918971
  • 30.05€ / Each
Estimated Shipment: 31-03-2025
Description
  • Highly durable.
  • Suitable for universal use.
  • Tote caddy can be used with 1 or 2 compartments.
Specifications
Specifications
Orange
385 mm
Utility Tray
2 long compartments
8.1 in.
Not Autoclavable
205 mm
HDPE
255 mm
15.2 in.
10 in.