Learn More
Lenz Laborglasinstrumente™ Tower Pycnometer
Brand:  Lenz Laborglasinstrumente™ 05660007
Product Code. 10748651
  • 129.00€ / Each
Estimated Shipment: 30-06-2025
Specifications
1 each
Tower Pycnometer
1000 mL
Tower Pycnometer
Product Title
Lenz Laborglasinstrumente™ Tower Pycnometer > Capacity: 10mL