Learn More
Thermo Scientific™ UHPLC Filters
Protects columns without compromising performance.
Brand:  Thermo Scientific™ 27006
276.25 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.
 View more versions of this product
Product Code. 10775706
  • 290.79€ / Each
Estimated Shipment: 05-05-2025
Description
Sacrificing performance is not an option with Thermo Scientific™ UHPLC filters that protect columns for Hypersil GOLD™ 1.9μm and Syncronis 1.7μm columns. These UHPLC filters provide a low-volume filter cartridge design, maintain peak shape, and ensure minimal efficiency loss through dispersion.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Each
UHPLC Direct Connect Filter Holder
Thermo Scientific
Product Title
Thermo Scientific™ UHPLC Filters >