Learn More
Thermo Scientific™ TraceGOLD TG-5SilMS GC Columns
Incorporates phenyl groups in the polymer backbone for improved thermal stability, reduced bleed and reduced susceptibility to oxidation.
334.00€ - 1385.00€
Specifications
Stationary Phase | TraceGOLD™ TG-5 SilMS |
---|---|
USP Type | G27, G36 |
Max. Temperature | 330/350°C |
For Use With (Application) | GC/MS Applications Using Ion-Trap Systems, GC/MS for Polycyclic Aromatics, Phthalates, Phenols, Amines, and Organophosphates, GC/MS for Hydrocarbons Including Chlorinated Hydrocarbons |
Polarity | Low-Polar |
Product Code | Brand | Length (Metric) | Diameter (Metric) Inner | Film Thickness | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Diameter (Metric) Inner | Film Thickness | Price | Quantity & Availability | |||||
11521174
|
Thermo Scientific™
26096-5800 |
40 m | 0.18 mm | 0.18 ÎĽm | N/A | ||||||
15297478
|
Thermo Scientific™
26096-5820 |
12 m | 0.2 mm | 0.33 ÎĽm | N/A | ||||||
10311595
|
Thermo Scientific™
26096-1300 |
15 m | 0.25 mm | 0.25 ÎĽm |
506.00€
Each |
Estimated Shipment: 10-07-2025
Log in to see stock. |
|||||
10794637
|
Thermo Scientific™
26096-1310 |
15 m | 0.32 mm | 0.25 ÎĽm | N/A | ||||||
10177894
|
Thermo Scientific™
26096-1420 |
30 m | 0.25 mm | 0.25 ÎĽm |
828.00€
Each |
In Stock
Log in to see stock. |
|||||
10457524
|
Thermo Scientific™
26096-1430 |
30 m | 0.32 mm | 0.25 ÎĽm |
862.00€
Each |
In Stock
Log in to see stock. |
|||||
10526005
|
Thermo Scientific™
26096-1540 |
60 m | 0.25 mm | 0.25 ÎĽm |
1370.00€
Each |
In Stock
Log in to see stock. |
|||||
10586375
|
Thermo Scientific™
26096-2110 |
15 m | 0.25 mm | 0.5 ÎĽm | N/A | ||||||
10239184
|
Thermo Scientific™
26096-2230 |
30 m | 0.25 mm | 0.5 ÎĽm |
825.00€
Each |
Estimated Shipment: 09-07-2025
Log in to see stock. |
|||||
10698905
|
Thermo Scientific™
26096-2240 |
30 m | 0.32 mm | 0.5 ÎĽm |
879.00€
Each |
Estimated Shipment: 08-07-2025
Log in to see stock. |
|||||
10260335
|
Thermo Scientific™
26096-2840 |
15 m | 0.25 mm | 1 ÎĽm | N/A | ||||||
10447914
|
Thermo Scientific™
26096-2960 |
30 m | 0.25 mm | 1 ÎĽm |
812.00€
Each |
Estimated Shipment: 04-07-2025
Log in to see stock. |
|||||
10046034
|
Thermo Scientific™
26096-2970 |
30 m | 0.32 mm | 1 ÎĽm |
873.00€
Each |
Estimated Shipment: 16-07-2025
Log in to see stock. |
|||||
10167664
|
Thermo Scientific™
26096-3080 |
60 m | 0.25 mm | 1 ÎĽm |
1385.00€
Each |
Estimated Shipment: 08-07-2025
Log in to see stock. |
|||||
10702347
|
Thermo Scientific™
26096-3360 |
30 m | 0.53 mm | 1.5 ÎĽm |
1010.00€
Each |
Estimated Shipment: 08-07-2025
Log in to see stock. |
|||||
13178295
|
Thermo Scientific™
26096-5780 |
20 m | 0.18 mm | 0.18 ÎĽm |
741.00€
Each |
Estimated Shipment: 11-07-2025
Log in to see stock. |
|||||
Description
Thermo Scientific™ TraceGOLD™ TG-5SilMS GC Columns incorporate phenyl groups in the polymer backbone for improved thermal stability, reduced bleed and reduced susceptibility to oxidation.
- Low polarity, silarylene phase with selectivity comparable to 5% diphenyl/95% dimethyl polysiloxane
- Designed for low bleed and outstanding inertness
- Similar to DB-5MS, VF-5ms, CP-Sil 8 Low-Bleed/MS, Rxi-5Sil MS, BPX5, ZB-5ms, Optima-5MS, SLB-5
Recommended for:
- GC/MS applications using ion-trap systems
- Polycyclic aromatics
- Hydrocarbons including chlorinated hydrocarbons
- Phthalates
- Phenols
- Amines
- Organophosphates
Specifications
TraceGOLD™ TG-5 SilMS | |
330/350°C | |
Low-Polar |
G27, G36 | |
GC/MS Applications Using Ion-Trap Systems, GC/MS for Polycyclic Aromatics, Phthalates, Phenols, Amines, and Organophosphates, GC/MS for Hydrocarbons Including Chlorinated Hydrocarbons |