missing translation for 'onlineSavingsMsg'
Learn More

trans-4-(Boc-aminomethyl)cyclohexanemethylamine, 97%, Thermo Scientific Chemicals

Product Code. 15427986
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Used as pharmaecutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as pharmaecutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 166168-16-7
Molecular Formula C13H26N2O2
Molecular Weight (g/mol) 242.363
MDL Number MFCD04112680
InChI Key NYXOBVVHJZENCO-UHFFFAOYSA-N
Synonym tert-butyl trans-4-aminomethyl cyclohexyl methyl carbamate, trans-l-4-boc-aminomethyl-cyclohexane methanamine, trans-l-4-boc-aminomethyl cyclohexane methanamine, trans-4-aminomethyl-cyclohexylmethyl-carbamic acid tert-butyl ester, trans-4-boc-aminomethyl cyclohexanemethanamine, tert-butyl n-4-aminomethyl cyclohexyl methyl carbamate, n-t-butyloxycarbonyl-1,4-trans-diaminomethyl-cyclohexane, carbamicacid, n-trans-4-aminomethyl cyclohexyl methyl-, 1,1-dimethylethyl ester
PubChem CID 2756046
IUPAC Name tert-butyl N-[[4-(aminomethyl)cyclohexyl]methyl]carbamate
SMILES CC(C)(C)OC(=O)NCC1CCC(CC1)CN

Specifications

Solubility Information Slightly soluble in water.
Formula Weight 242.35
Percent Purity 97%
Quantity 250 mg
Chemical Name or Material trans-4-(Boc-aminomethyl)cyclohexanemethylamine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'