Learn More
enQuireBio™ TranslationBlocker Human ApoE siRNA
Blocks transcription and translation of ApoE from Human.
Brand: enQuireBio™ QX5-2nmol
This item is not returnable.
View return policy
Specifications
NP_000032 | |
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. | |
348 | |
2 nmol | |
Human |
Ovcar3 Human Ovarian Cancer Cells. | |
APOE | |
siRNA | |
AD2, APO-E, LPG, LDLCQ5, ApoE4, ApoE, Apolipoprotein E | |
Lyophilized |