missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ TranslationBlocker Human Caspase-7 siRNA

Blocks transcription and translation of Caspase-7 from Human.

250.00€ - 369.00€

Specifications

Accession Number NP_001218
Cell Lines Tested 293 Human Embryonic Kidney Cells with NF-kappa B and Flag tagged DR4 constructs.
Content And Storage Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month.
Gene Symbol CASP7
Gene ID (Entrez) 840
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16059231
View Documents
enQuireBio™
QX42-2nmol
2 nmol This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A
16049231
View Documents
enQuireBio™
QX42-10nmol
10 nmol This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A
Description

Description

Specifications

Specifications

NP_001218
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month.
840
ICE-LAP3, CMH-1, CASP-7, MCH3, LICE2, CASP7
Lyophilized
293 Human Embryonic Kidney Cells with NF-kappa B and Flag tagged DR4 constructs.
CASP7
siRNA
Human
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : This product is guaranteed to work for a period of two years when stored at -70°C or colder, and one year when aliquoted and stored at -20°C.

SDS
Documents

Documents

Product Certifications