Learn More
enQuireBio™ TranslationBlocker Human Cellular tumor antigen p53 siRNA
Blocks transcription and translation of Cellular tumor antigen p53 from Human.
Brand: enQuireBio™ QX32-2nmol
This item is not returnable.
View return policy
Specifications
NP_000537 | |
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. | |
7157 | |
2 nmol | |
Human |
U-2 OS Huan Osteosarcoma Epithelial Cells from Bone. | |
TP53 | |
siRNA | |
TRP53, BCC7, P53, LFS1, TP53 | |
Lyophilized |