missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ TranslationBlocker Human ErbB2 / HER2 siRNA
Blocks transcription and translation of ErbB2 / HER2 from Human.
Brand: enQuireBio™ QX19-10nmol
This item is not returnable.
View return policy
Specifications
NP_004439 | |
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. | |
2064 | |
10 nmol | |
Human |
BT-474 Human Breast Carcinoma Cells, SkBr3 Human Breast Adenocarcinoma Cells, and MCF7 Human Breast Adenocarcinoma Cells. | |
ERBB2 | |
siRNA | |
CD340, NGL, HER-2/neu, HER2, MLN 19, NEU, HER-2, TKR1, Receptor tyrosine-protein kinase erbB-2 | |
Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction