missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ TranslationBlocker Human IRAK4 siRNA
Blocks transcription and translation of IRAK4 from Human.
Brand: enQuireBio™ QX29-10nmol
This item is not returnable.
View return policy
Specifications
NP_057207 | |
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. | |
51135 | |
10 nmol | |
Human |
293T Human Embryonic Kidney Cells with Large T Antigen. | |
IRAK4 | |
siRNA | |
Interleukin-1 receptor-associated kinase 4, IRAK-4 | |
Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction