Learn More
enQuireBio™ TranslationBlocker Human IRAK4 siRNA
Blocks transcription and translation of IRAK4 from Human.
Brand:  enQuireBio™ QX29-2nmol
 View more versions of this product
Product Code. 16059201
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Explore more special offers
Specifications
NP_057207
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month.
51135
2 nmol
Human
293T Human Embryonic Kidney Cells with Large T Antigen.
IRAK4
siRNA
Interleukin-1 receptor-associated kinase 4, IRAK-4
Lyophilized