Learn More
enQuireBio™ TranslationBlocker Human TLR4 siRNA
Blocks transcription and translation of TLR4 from Human.
Brand:  enQuireBio™ QX30-2nmol
 View more versions of this product
Product Code. 16099201
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Explore more special offers
Specifications
NP_612564
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month.
7099
2 nmol
Human
HepG2 Human Carcinoma Cells from Liver.
TLR4
siRNA
TLR-4, Toll-like Receptor 4, CD284
Lyophilized