missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ TranslationBlocker Human TLR9 / CD289 siRNA
Blocks transcription and translation of TLR9 / CD289 from Human.
250.00€ - 369.00€
Specifications
Accession Number | NP_059138 |
---|---|
Cell Lines Tested | 293T Human Embryonic Kidney Cells with Large T Antigen. |
Content And Storage | Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. |
Gene Symbol | TLR9 |
Gene ID (Entrez) | 54106 |
Specifications
NP_059138 | |
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. | |
54106 | |
CD289, Toll-like receptor 9, TLR-9 | |
Lyophilized |
293T Human Embryonic Kidney Cells with Large T Antigen. | |
TLR9 | |
siRNA | |
Human |
Safety and Handling
missing translation for 'shelfLife' : This product is guaranteed to work for a period of two years when stored at -70°C or colder, and one year when aliquoted and stored at -20°C.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title