Learn More
enQuireBio™ TranslationBlocker Mouse TNF alpha siRNA
Blocks transcription and translation of TNF alpha from Mouse.
Brand:  enQuireBio™ QX23-10nmol
 View more versions of this product
Product Code. 16029191
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Explore more special offers
Specifications
NP_038721
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month.
21926
10 nmol
Mouse
Murine Peritoneal Macrophage.
Tnf
siRNA
DIF, TNFA, TNFSF2, TNLG1F, TNF-alpha, Tumor necrosis factor alpha, Tumor necrosis factor
Lyophilized