Learn More
enQuireBio™ TranslationBlocker Mouse TNF alpha siRNA
Blocks transcription and translation of TNF alpha from Mouse.
Brand: enQuireBio™ QX23-2nmol
This item is not returnable.
View return policy
Specifications
NP_038721 | |
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. | |
21926 | |
2 nmol | |
Mouse |
Murine Peritoneal Macrophage. | |
Tnf | |
siRNA | |
DIF, TNFA, TNFSF2, TNLG1F, TNF-alpha, Tumor necrosis factor alpha, Tumor necrosis factor | |
Lyophilized |