missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ TranslationBlocker Rat Estrogen receptor beta siRNA
Blocks transcription and translation of Estrogen receptor beta from Rat.
250.00€ - 369.00€
Specifications
Accession Number | NP_036886 |
---|---|
Cell Lines Tested | Rat Primary Osteoblasts. |
Content And Storage | Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. |
Gene Symbol | Esr2 |
Gene ID (Entrez) | 25149 |
Specifications
NP_036886 | |
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. | |
25149 | |
ERbeta, Erb2, ER-beta | |
Lyophilized |
Rat Primary Osteoblasts. | |
Esr2 | |
siRNA | |
Rat |
Safety and Handling
missing translation for 'shelfLife' : This product is guaranteed to work for a period of two years when stored at -70°C or colder, and one year when aliquoted and stored at -20°C.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ TranslationBlocker Rat Estrogen receptor beta siRNA