missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ TranslationBlocker Rat Estrogen receptor siRNA
Blocks transcription and translation of Estrogen receptor from Rat.
Brand: enQuireBio™ QX15-2nmol
This item is not returnable.
View return policy
Specifications
NP_036821 | |
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. | |
24890 | |
2 nmol | |
Rat |
Rat Primary Osteoblasts. | |
Esr1 | |
siRNA | |
RNESTROR, ER-alpha, Esr, ESRA, ERA, ERAlpha, ER | |
Lyophilized |