Learn More
Fisherbrand™ Translucent Silicone Tubing
Use this tubing with peristaltic pumps. Fisherbrand™ Translucent Silicone Tubing is ideal for pharmaceutical, biotechnology and sterile fluid transfer applications.
Brand: Fisherbrand™ 1174-3416
This item is not returnable.
View return policy
Description
- Translucent silicone tubing
- FDA compliant (food)
Specifications
FDA, BFR and WRAS approved | |
Silicone | |
25 m | |
Peristaltic pumps | |
-40°C | |
Translucent | |
250°C |
7 mm | |
10 mm | |
1.5 mm | |
Pharmaceutical , biotechnology, transfer sterile fluids | |
Sterile | |
Flexible |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 13
Product Content Correction
Product Title
Fisherbrand™ Translucent Silicone Tubing > I.D.: 7mm; O.D.: 10mm; Length: 25m