Learn More
Fisherbrand™ Translucent Silicone Tubing
Use this tubing with peristaltic pumps. Fisherbrand™ Translucent Silicone Tubing is ideal for pharmaceutical, biotechnology and sterile fluid transfer applications.
Brand: Fisherbrand™ 1178-3855
Description
- Translucent silicone tubing
- FDA compliant (food)
Specifications
FDA, BFR and WRAS approved | |
Silicone | |
5 m | |
Peristaltic pumps | |
-40°C | |
Translucent | |
250°C |
5 mm | |
8 mm | |
1.5 mm | |
Pharmaceutical , biotechnology, transfer sterile fluids | |
Sterile | |
Flexible |