Learn More
Zeiss™ Transmitted Light Equipment S
Ergonomic console with light guide and two adjustable reflectors for bright field/dark field and for variable oblique light
Brand:  Zeiss™ 435501
Product Code. 13592330
  • 2780.00€ / Each
Estimated Shipment: 05-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Compatible Microscopes:
  • Axio Zoom.V16
  • SteREO Discovery.V12
  • SteREO Discovery.V20
  • SteREO Discovery.V8
  • SteREO Lumar.V12
Specifications
Specifications
Transmitted Light Equipment
Transmitted Light
Light guide and 2 shiftable and tiltable reflectors for bright field/dark field and for variable oblique light
Transmitted light equipment
Stand Base 450