Learn More
VITLAB™ Transparent PFA Narrow-Mouth Bottle with Buttress Threads
VITLAB™ Transparent PFA Narrow-Mouth Bottle with Buttress Thread are supplied with a screw cap. Made of PFA, they are ideal for long-term storage of high-purity oxidants, acids, and alkalis, as well as hydrocarbons, trace analysis solvents, and standards.
Brand: VITLAB™ 108297
This item is not returnable.
View return policy
Description
- Product Type: Storage Bottle
- Description: Narrow Mouth with Buttress Thread
- Material: PFA
- Shape: Round
- Includes: Screw Cap
Specifications
Anti-adhesive and Smooth | |
Transparent | |
Saving and Storing | |
PFA | |
1Each | |
-200°C to +260°C | |
Autoclavable | |
No | |
Round | |
Narrow Mouth Bottle | |
PFA | |
61 mm |
250 mL | |
2.4 in. | |
Screw cap | |
-328°F to +500°F | |
Non-sterile | |
Yes | |
Reusable | |
Narrow | |
160 mm | |
6.3 in. | |
S 28 | |
Screw Cap |
Product Content Correction
Product Title
VITLAB™ Transparent PFA Narrow-Mouth Bottle with Buttress Threads > Capacity: 250 mL, Diameter: 61 mm, Height: 160 mm