Learn More
Kartell™ Transparent Polypropylene Powder Funnels
BIOPHARMA
Manufactured with polypropylene for strength and durability. Kartell™ Transparent Polypropylene Powder Funnels are designed to funnel large quantities of powders or other solid or liquid materials without flow rate restrictions, powder segregation or bridging.
Brand:  Kartell™ 0017100
Product Code. 11954299
  • 29.80€ / Pack of 20
Estimated Shipment: 20-05-2025
Description
Manufactured with polypropylene for strength and durability. Kartell™ Transparent Polypropylene Powder Funnels are designed to funnel large quantities of powders or other solid or liquid materials without flow rate restrictions, powder segregation or bridging.
  • 60° angled sides
  • External ribbing avoids pressure lock
  • Can be used with liquids
  • Autoclavable
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberT23S01397 Certificate TypeDeclaration of Conformity Date03/04/2025 Catalog Number0017100
Lot NumberT24I02978 POWDER Certificate TypeDeclaration of Conformity Date12/12/2024 Catalog Number0017100
Lot NumberT24I02978 Certificate TypeDeclaration of Conformity Date15/11/2024 Catalog Number0017100
Lot NumberT23S01397 Certificate TypeDeclaration of Conformity Date01/08/2024 Catalog Number0017100
Lot Number T23S01397 Certificate TypeDeclaration of Conformity Date06/06/2024 Catalog Number0017100
Product Title
Kartell™ Transparent Polypropylene Powder Funnels > Capacity: 45mL; Length: 45mm