Learn More
uvex Trend Black/Blue PU/PU 69342 Low Shoe - Width 11
Uvex 2 Trend safety footwear for medium-risk applications (S1-S3); heel cap offers increased stability without limiting movement
107.00€
Specifications
Color | Black, Blue |
---|---|
Material | Leather |
Sole Material | Dual-density polyurethane (PU/PU) |
Toe Material | Steel |
Disposable | Reusable |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
17243591
|
uvex
6934238 |
38 |
107.00€
1 pair |
Estimated Shipment: 28-01-2025
Log in to see stock. |
|||||
17253591
|
uvex
6934239 |
39 |
107.00€
1 pair |
Estimated Shipment: 28-01-2025
Log in to see stock. |
|||||
17263591
|
uvex
6934240 |
40 |
107.00€
1 pair |
Estimated Shipment: 28-01-2025
Log in to see stock. |
|||||
17273591
|
uvex
6934241 |
41 |
107.00€
1 pair |
Estimated Shipment: 28-01-2025
Log in to see stock. |
|||||
17283591
|
uvex
6934242 |
42 |
107.00€
1 pair |
Estimated Shipment: 28-01-2025
Log in to see stock. |
|||||
17293591
|
uvex
6934244 |
44 |
107.00€
1 pair |
Estimated Shipment: 28-01-2025
Log in to see stock. |
|||||
17203601
|
uvex
6934245 |
45 |
107.00€
1 pair |
Estimated Shipment: 28-01-2025
Log in to see stock. |
|||||
17213601
|
uvex
6934246 |
46 |
107.00€
1 pair |
Estimated Shipment: 28-01-2025
Log in to see stock. |
|||||
17223601
|
uvex
6934247 |
47 |
107.00€
1 pair |
Estimated Shipment: 28-01-2025
Log in to see stock. |
|||||
17233601
|
uvex
6934248 |
48 |
107.00€
1 pair |
Estimated Shipment: 28-01-2025
Log in to see stock. |
|||||
17243601
|
uvex
6934249 |
49 |
107.00€
1 pair |
Estimated Shipment: 28-01-2025
Log in to see stock. |
|||||
17253601
|
uvex
6934250 |
50 |
107.00€
1 pair |
Estimated Shipment: 28-01-2025
Log in to see stock. |
|||||
17263601
|
uvex
6934251 |
51 |
107.00€
1 pair |
Estimated Shipment: 28-01-2025
Log in to see stock. |
|||||
17273601
|
uvex
6934252 |
52 |
107.00€
1 pair |
Estimated Shipment: 28-01-2025
Log in to see stock. |
|||||
Description
Uvex 2 Trend Shoe 69342 S3 SRC
- Sporty S3 safety shoe with wide fit
- All sole materials free of silicones, plasticizers and other paint wetting impairment substances
- Smooth-running, dirt and heat-resistant lacing system
- Meets ESD specification with volume resistance under 35 megohms
- Reliable protection for wearers
- Penetration-resistant steel midsole, steel toecap and stabilizing Uvex anti-twist heel cap to prevent ankle twisting
- Ergonomically designed outsole made from dual-density polyurethane with excellent slip resistance
- Reflective details for enhanced visibility
- Foamed polyurethane scuffcap helps increase product life of upper
- Uvex Climazone technology lining, upper and removable comfortable climatic insole
- Moisture transport system helps maintain foot temperature
- Soft padding collar and dust tongue
- Removable and anti-static comfortable insole with a moisture transport system and additional shock absorption around heel and forefoot
- Article No. 69342
- Standard EN ISO 20345:2011 S3 SRC
- Sizes: 38 to 52
Specifications
Black, Blue | |
Dual-density polyurethane (PU/PU) | |
Reusable | |
Unisex | |
Uvex 2 | |
No | |
SRC |
Leather | |
Steel | |
EN ISO 20345:2011 | |
Standard Width | |
Yes | |
S3 | |
ESD Safety Shoe |