missing translation for 'onlineSavingsMsg'
Learn More
Trichloroethylene, 99+%, extra pure, stabilized, Thermo Scientific™ Product Code.: 10766581

Trichloroethylene, 99+%, extra pure, stabilized, Thermo Scientific™

Product Code. 10766581 Shop All Thermo Scientific Chemicals Products
1L; Glass bottle
Click to view available options
1L; Glass bottle
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 10766581

Brand: Thermo Scientific Acros 158310010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0471563 Certificate TypeCertificate of Analysis Date17/05/2025 Catalog Number158310025,158310010,158310250
Lot NumberA0471563 Certificate TypeCertificate of Analysis Date03/04/2025 Catalog Number158310010,158310250
Lot NumberA0469246 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number158310025,158310010,158310250
Lot NumberA0460455 Certificate TypeCertificate of Analysis Date21/02/2024 Catalog Number158310025,158310010,158310250
Lot NumberA0459654 Certificate TypeCertificate of Analysis Date25/01/2024 Catalog Number158310025,158310010,158310250
compliance-icons compliance-icons
Product Identifier
  • Trichloroethylene, stabilized
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • H341-Suspected of causing genetic defects.
  • H350-May cause cancer.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Trichloroethylene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Trichloroethylene, 99+%, extra pure, stabilized, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.