Learn More
Thermo Scientific™ TriPlus RSH Autosampler Fixed GT Needle Syringe
Thermo Scientific™ TriPlus RSH Autosampler Fixed GT Needle Syringes are accurate and reproducible over a range of volumes.
Brand: Thermo Scientific™ 365I2351
200.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Thermo Scientific™ TriPlus RSH Autosampler Fixed GT Needle Syringes are accurate and reproducible over a range of volumes.
- Borosilicate glass with metal flanges
- Needles and plungers are stainless steel
- Field-proven syringe design
Specifications
Stainless Steel (Needle), Stainless Steel (Plunger) | |
23 G | |
GC Autosampler Syringe | |
250 μL | |
Side Hole | |
Fixed | |
Yes |
250 μL FN GT, 23 G, 57 mm | |
57 mm | |
Ea. | |
57 mm | |
TriPlus RSH Autosampler | |
Sterile |