missing translation for 'onlineSavingsMsg'
Learn More

TRIS-acetate-SDS running buffer (10X), pH 8.3, Thermo Scientific™

53.27€ - 80.10€

Specifications

Chemical Name or Material TRIS-acetate-SDS running buffer (10X)
Physical Form Liquid
Packaging Plastic bottle
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15414919
View Documents
Thermo Scientific Alfa Aesar
J62560.AP
500 mL This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A
15424919
View Documents
Thermo Scientific Alfa Aesar
J62560.K2
1 L This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

TRIS-acetate-SDS running buffer (10X)
Liquid
Plastic bottle
Videos
Safety and Handling

Safety and Handling

H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
TRIS-acetate-SDS running buffer (10X), pH 8.3, Thermo Scientific™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.