missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris-Glycine-SDS, 1X Solution (Electrophoresis), Fisher BioReagents
Brand: Fisher Bioreagents BP2440-4
This item is not returnable.
View return policy
Description
Tris-Glycine-SDS is commonly used as a buffer in SDS-PAGE.Specifications
Tris-Glycine-SDS | |
56-40-6, 77-86-1, 151-21-3, 7732-18-5 | |
4 L | |
Filtered through a 0.2-micron filter. | |
Not Detected |
1X Solution | |
8.3 to 8.5 (25°C) | |
Not Detected | |
Poly CUBE | |
Electrophoresis |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction