missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tuberin/TSC2 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68380-1-IG-20UL
This item is not returnable.
View return policy
Description
The Tuberin/TSC2 antibody from Proteintech is a mouse monoclonal antibody generated with peptide of human Tuberin/TSC2. This antibody recognizes human, mouse, rat antigen. The Tuberin/TSC2 antibody has been validated for the following applications: WB, IHC, IF/ICC, ELISA analysis.Specifications
Tuberin/TSC2 | |
Monoclonal | |
1000 μg/mL | |
FLJ43106; LAM; TSC4 | |
Peptide | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
200 kDa | |
Liquid |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, ELISA | |
1A10B2 | |
Unconjugated | |
Mouse | |
Protein A purification | |
RUO | |
7249 | |
Store at -20°C. Stable for one year after shipment. | |
Monoclonal Antibody | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Tuberin/TSC2 Mouse Monoclonal Antibody, Proteintech > Unconjugated; 20 μL
Spot an opportunity for improvement?Share a Content Correction