Learn More
Kartell™ Tapered Hose Coupling
Polyethylene disconnectors provide a quick, flexible, and safe alternative to expensive adapters. Kartell™ Tapered Hose Couplings tightly join glass, rubber, or plastic hoses together.
Brand: Kartell™ 7043400
This item is not returnable.
View return policy
Description
- Useful for joining hoses of glass, rubber or plastic
- Connector fits tightly together
- Replaces expensive adapters
Specifications
CE/IVD | |
Bore Size 1.6 mm | |
4/5/6 mm | |
Glass, rubber, or plastic hoses | |
1.6 mm |
4 to 6 mm | |
Polyethylene | |
Hose Coupling | |
10 | |
Natural |