Learn More
Saint Gobain Life Sciences Versilon™ GSR Tubing
Constructed of 45 Shore A natural rubber. Saint Gobain Life Sciences Versilon™ GSR Tubing is light, supple and wear-resistant with good impermeability and provides an ideal combination of chemical resistance and pump life.
Brand:  Saint Gobain Life Sciences 710064
Product Code. 11751545
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Description
Description
  • 45 shore A natural rubber construction
  • Autoclavable
  • Good chemical resistance to acids and bases
  • Very good elasticity and flexibility
  • Good resistance to abrasion
Applications include:
  • Fluid handling in food & beverage industry
  • Semi-vacuum application
  • Vacuum application
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Autoclavable
9 mm
2.5 mm
Red
4 mm
25 m
-40°C
70°C