Learn More
SCHOTT AG Lighting and Imaging™ Tubular diffuser Only for VisiLED Ring Lights
Tubular diffuser designed for use with LED ringlights from EasyLED or VisiLED series for stereomicroscopy illumination.
Brand:  SCHOTT AG Lighting and Imaging™ 400590
Product Code. 17853438
  • 269.00€ / Pack of 20
Estimated Shipment: 03-01-2025
Description
VisiLED diffuser made of improved materials is designed specifically for use only with VisiLED brightfield ring lights (S80-65N-2).
  • Prevents LED reflections
  • High transmission and perfect haze of more than 99%
  • Includes adapting thread (M90 x 1)
  • Controller needs to be ordered separately
Specifications
Specifications
Tubular Diffuser
A Wide Variety of Tasks From Life Science to Industrial Applications
EasyLED Ring light (600 200), EasyLED Ringlight Plus (600 300), VisiLED range S80-25 and S80-55 Ringlights (400225 and 400255) manufactured from August 2023