Learn More
Rogo Sampaic™ Tungsten Carbide Anatomical Heavy Duty Scissors
124.00€ - 147.00€
Specifications
Product Type | Heavy Duty Anatomical Scissors |
---|---|
Length (Metric) | 140 mm |
Material | Tungsten Carbide |
Product Code | Brand | Tip Style | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Tip Style | Price | Quantity & Availability | ||||||
11726145
|
Rogo Sampaic™
FMN021 |
Sharp/Sharp |
127.00€
Each |
Estimated Shipment: 03-06-2025
Log in to see stock. |
||||||
11736145
|
Rogo Sampaic™
FMN031 |
Round/Round |
147.00€
Each |
In Stock
Log in to see stock. ![]() |
||||||
11716145
|
Rogo Sampaic™
FMN022 |
Sharp/Round |
124.00€
Each |
Estimated Shipment: 24-04-2025
Log in to see stock. |
||||||
Specifications
Heavy Duty Anatomical Scissors | |
140 mm | |
Tungsten Carbide |
Product Title
Rogo Sampaic™ Tungsten Carbide Anatomical Heavy Duty Scissors