Learn More
RETSCH Tungsten Carbide Grinding Jars
Use these grinding jars for dry, wet and cryogenic grinding of small samples. Retsch™ Tungsten Carbide Grinding Jars feature a self-centering mechanism and a screw-top design for leak-proof grinding.
Brand: RETSCH 01.462.0115
This item is not returnable.
View return policy
Description
- Screw-top Tungsten grinding jars
- Available in a variety of sizes
- For use with Retsch mills
Specifications
Grinding Jar | |
5 mL | |
For MM 200 Mixer Mill | |
Tungsten Carbide |