Learn More
Brannan™ Twaddell Hydrometers
For reporting the measured specific gravity of a liquid relative to water. Brannan™ Twaddell Hydrometers are manufactured from soda glass, steel shot ballast and synthetic wax resin.
Brand:  Brannan™ 10/104/0
Additional Details : Weight : 0.20000kg
 View more versions of this product
Product Code. 12859177
  • 13.55€ / Each
Estimated Shipment: 13-08-2024
Description
For reporting the measured specific gravity of a liquid relative to water. Brannan™ Twaddell Hydrometers are manufactured from soda glass, steel shot ballast and synthetic wax resin.
  • 160mm in length
  • With single degree divisions
  • Adjusted for use at 15.6°C (60°F)
  • For medium surface tension liquids
  • Available for numerous Twaddell ranges
Specifications
Specifications
1Pack
±1Tw
160 mm
74 to 102Tw
Twaddle Hydrometer
1Tw
Steel Shot