Learn More
Thermo Scientific Chemicals Tween 20 washing buffer, 1.0% in PBS (20X)
126.63€ - 226.13€
Specifications
Product Type | Buffer |
---|---|
Form | Liquid |
Concentration | 1.0% in PBS (20X) |
Content And Storage | Ambient temperatures |
Description
It is used as wash buffers for ELISA, Western blot and other immunoassays as well as a blocking buffer for plate-based assays.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Buffer | |
Liquid | |
1.0% in PBS (20X) | |
Ambient temperatures |