Documents
Product Information
Manuals
- 1
- 2
Support a wide range of sample sizes and applications with the swinging bucket rotor.
Brand: Thermo Scientific™ 75003656
422.94 EUR valid until 2025-04-30
Use promo code "24155" to get your promotional price.
Support a wide range of sample sizes and applications with the Thermo Scientific™ TX-400 4 x 400mL Swinging Bucket Rotor.
Ordering Information:
Buckets and lids are sold separately; order rotor body (75-003-629) and components to complete assembly. For replacement O-rings for lids (set of four), order 75-003-657. Tube adapters are also available separately. Contact your sales representative for information.
Compatible with:
Thermo Scientific Sorvall™ ST16, SL16, Sorvall Legend™ X1, Multifuge™ X1, Megafuge™ 16 Centrifuge Series
Recommended for:
Cell culture, blood separation, body fluids processing, microbiology, proteomics, genomics and molecular biology
ClickSeal Biocontainment Lids | |
Thermo Scientific Sorvall Legend X1, Sorvall ST 16, Multifuge X1, Megafuge 16 and SL 16 Centrifuges | |
Yes |
Bucket Lid | |
Biocontainment certified by CAMR™, HPA, Porton Down, UK | |
Set of 4 |
Expires: 2025-12-31
Expires: 2025-05-01