missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals [Tyr0] Gastric Inhibitory Peptide (23-42), Human

For cellular and molecular biology applications

164.82€ - 316.00€

Specifications

Species Human
Purification Method HPLC
Common Name [Tyr22] Gastric Inhibitory Peptide (22-42), human
Content And Storage -30°C to -10°C
Format Solid
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15420507
View Documents
Thermo Scientific Chemicals
0.5 mg
164.82€
0.50mg
Estimated Shipment: 16-12-2024
to see stock.
Add to basket
 
15457349
View Documents
Thermo Scientific Chemicals
J66109.M^
1 mg This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Human
[Tyr22] Gastric Inhibitory Peptide (22-42), human
Solid
[Tyr0] Gastric Inhibitory Peptide (23-42), human
Tyr-Val-Asn-Trp-Leu-Leu-Ala-Gln-Lys-Gly-Lys-Lys-Asn-Asp-Trp-Lys-His-Asn-Ile-Thr-Gln
HPLC
-30°C to -10°C
2584.90
RUO - research use only
Videos
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only