Learn More
DuPont™ Tyvek 500 lab cot wuth zipper
Offers enhanced protection for body parts that are more exposed to hazardous substances. DuPont™ Tyvek™ 500 Lab Coat with Zipper and Pockets are composed of flash spun high density polyethylene, providing an idea balance of protection, durability and comfort.
Brand: DuPont™ D13495983
286.92 EUR valid until 2025-04-30
Use promo code "24155" to get your promotional price.
Description
- Silicon non-added
- With collar, zipper closure and 2 pockets
- Permeable to both air and water vapour, yet repels water-based liquids and aerosols
- Excellent barrier against fine particles and fibres (down to 1 micron in size), is ultra-low-linting and antistatically treated
- Applications include pharmaceutical handling, chemical protection, lead and asbestos abatement/remediation, general maintenance/operations, spray painting and general clean-up
Applications and solutions series: Safety edition
File size: 7.7 MB
Product Content Correction
Product Title
DuPont™ Tyvek 500 lab cot wuth zipper > Size: Medium