Learn More
DuPont™ Tyvek™ 600 Hooded Coverall, Plus Size
Shop All DuPont Products
Click to view available options
Size:
4X-Large
5X-Large
6X-Large
7X-Large
Description
- Thumb loops
- Stitched and over-taped seams for protection and strength
- Tyvek™ zipper and zipper flap for enhanced protection
- Face, wrist and ankle tunnelled elastication, for good fit and contamination/ linting reduction
Specifications
Specifications
Size | 4X-Large |
Color | White |
Closure Type | Zipper |
Seam Type | Stitched, Taped |
Certifications/Compliance (Europe) | (EU) 2016/425, EN 14126, EN 1149-5 |
Product Content Correction
Product Title
DuPont™ Tyvek™ 600 Hooded Coverall, Plus Size >