Learn More
DuPont™ Tyvek 500 Jacket
Shop All DuPont Products
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
Description
- Specially designed for use with Tyvek™ apparel
- Flash-spun HDPE construction
- Stitched internal seams
- Front zip closure
- Elasticated face and wrists
- White
- PPE Category I
- Antistatic treatment (EN 1149-5) - on both sides
Specifications
Specifications
Size | X-Large |
Material | HDPE |
Color | White |
Closure Type | Zipper |
Certifications/Compliance | EN 1149-5 |
Seam Type | Stitched (Internal) |
Product Type | Hooded Chemical-resistant Jacket |
For Use With (Application) | Pharmaceutical Handling, Chemical Protection, Lead and Asbestos Abatement/remediation, General Maintenance/operations, Spray Painting and General Clean-up, Amongst Many Others |
Product Suggestions
Customers who viewed this item also viewed.
Special Offers
-
Multiple Manufacturers
Gear Up for Safety
Save up to 60% on gloves, apparel, and morePromo Code: 24131Expires: 2025-06-08
Product Content Correction
Product Title
DuPont™ Tyvek 500 Jacket >