Learn More
DuPont™ Tyvek™ IsoClean™ Hood with Attached Mask
Click to view available options
Product Type: Hood
Description
- Bound face opening for low particle shedding
- Aseptically folded
- Breathable and low linting
- The accessories are individually double packed
- The attached mask is a blue sterile cleanroom mask made out of pleated polyethylene outer layer having a width of 17.5 cm. Please note that it is not a PPE (e.g. not an FFP1, 2 or 3) mask.
Specifications
Specifications
Product Type | Hood |
Size | One Size |
Closure Type | Ties with Loops |
Model | IC 689 |
Seam Type | Bound |
Sterility | Sterile |
Certifications/Compliance (Europe) | ISO 11137-1, ISO Class 5 |
For Use With (Application) | Cleanrooms Within The Biotech, Pharmaceutical, Medical Device Manufacturing, Food Processing, Cosmetics Industry as Well as in Other Critical or Controlled Environments. |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
4 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
AHS2308143 | Certificate of Sterility | 28/04/2023 | D15553188 |
AHS2244158 | Certificate of Sterility | 07/12/2022 | D15553188 |
AHS2239140 | Certificate of Sterility | 26/10/2022 | D15553188 |
AHS2235147 | Certificate of Sterility | 28/09/2022 | D15553188 |
Product Content Correction
Product Title
DuPont™ Tyvek™ IsoClean™ Hood with Attached Mask >