Learn More
DuPont™ Tyvek™ IsoClean™ IC 270B WS Lab Coats, Sterile, Double Bagged
Description
- Clean-processed and gamma-sterilized
- Bound internal seams
- Tunnelled elastication at wrists
- Front snap closure for easy donning and doffing
Tyvek™ IsoClean™ delivers an ideal balance of protection, durability and comfort. Made of high density polyethylene using a patented flash spinning process Tyvek™ IsoClean™ provides an inherent barrier to particles, microorganisms and non-hazardous light liquid splash. Tyvek™ IsoClean™ is also breathable and low linting.
Tyvek™ IsoClean™ accessories (option code WS) have been sterilized by gamma- irradiation. They are folded to aid aseptic donning and packaged in an ISO class 5 cleanroom. All DuPont™ Tyvek™ IsoClean™ sterile accessories (option code WS) are packed in a dual barrier packaging system, consisting of an inner and outer easy tear cleanroom bag. The packaging system serves as a key element for contamination risk reduction when transferring apparel into clean areas. The accessories are individually packed and grouped together in an outer bag.
Garments and accessories made of sterile Tyvek™ IsoClean™ are typically used in cleanrooms within the hospital pharmacies, biotech, pharmaceutical, medical device manufacturing, food processing, cosmetics industry as well as in other critical or controlled environments.
Specifications
Specifications
Product Type | Controlled Environments Lab Coat |
Size | Large |
Gender | Unisex |
Material | Tyvek |
Color | White |
Closure Type | Snaps |
Cuff Style | Elastic |
Disposable | Single-use |
Sterility | Sterile |
Resistance | Particulates, Light Liquid Splash |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Documents
Scientific Resources
Brochures
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
AHS2435132 | Certificate of Sterility | 11/12/2024 | D15560761 |
AHS2417133 | Certificate of Sterility | 20/07/2024 | D15560761 |
AHS2425145 | Certificate of Sterility | 20/07/2024 | D15560761 |
AHS2413115 | Certificate of Sterility | 20/07/2024 | D15560761 |
AHS2234134 | Certificate of Sterility | 24/05/2024 | D15560761 |
Special Offers
-
Multiple Manufacturers
Cleanroom Essentials Extravaganza
Gear Up Your Controlled EnvironmentPromo Code: 24151Expires: 2025-05-02
New Offer Save Up to 92%