missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TYW3 Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00€ - 399.00€
Specifications
Antigen | TYW3 |
---|---|
Concentration | 500 μg/mL |
Applications | Western Blot, ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17211436
|
Proteintech
29213-1-AP-20UL |
20 μL |
140.00€
20µL |
Estimated Shipment: 24-09-2024
Log in to see stock. |
|||||
17201436
|
Proteintech
29213-1-AP-150UL |
150 μL |
399.00€
150µL |
In Stock
Log in to see stock. |
|||||
Description
The TYW3 antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human TYW3. This antibody recognizes human antigen. The TYW3 antibody has been validated for the following applications: WB,ELISA analysis.
Specifications
TYW3 | |
Western Blot, ELISA | |
Unconjugated | |
Rabbit | |
127253 |
500 μg/mL | |
Polyclonal | |
Liquid | |
Human | |
Primary |